Would you like to log in again?
Cole-Parmer® Large Capacity Digital Hotplates, Antylia Scientific
Supplier: Antylia Scientific
Cole-Parmer®
04807-67 04807-83 04807-81
76539-134EA 1059.52 USD InStock
76539-134 76539-122 76539-136
Cole-Parmer® Large Capacity Digital Hotplates, Antylia Scientific
Heaters Hot Plates
Digital, microprocessor control for high-performance. Available with ceramic or aluminum plate.
  • Accurate digital setting and control of plate temperature
  • Be safe -flashing light warns when top plate is hot
  • Plate made from ceramic or aluminum/silicon alloy
These large capacity digital hotplates provide uniform heating conditions, and the microprocessor control allows for accurate plate temperature monitoring. The housing is shaped for stability deflecting spills away from the user. The ‘Hot’ warning light will flash whenever the plate temperature is above 50 °C. Accommodates multiple vessels, one large container, or even microscope slides.
Choice of top plate material:
Glass ceramic: provides excellent chemical resistance, high plate temperatures, easy to clean
Aluminum/silicon alloy – rapid heating and even temperature distribution across whole plate
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty