Would you like to log in again?
Cole-Parmer® Large Capacity Analog Infrared Hotplate Stirrer, Antylia Scientific
Supplier: Antylia Scientific
04807-87
76539-140EA 1403.41 USD InStock
76539-140
Cole-Parmer® Large Capacity Analog Infrared Hotplate Stirrer, Antylia Scientific
Stirrers Magnetic Hotplate Stirrers
Easy to use control dials.
  • Infrared heater is efficient and saves time and energy
  • Ideal for heating and stirring large volumes of liquid
  • Chemically resistant ceramic top
This hotplate stirrer features an efficient, 900-watt infrared heater that can boil a liter of water over 30% faster than a conventional, 1200-watt ceramic hotplate. The chemical-resistant, ceramic top plate has a 5.5" (14 cm) diameter heating area that stays cool around the edges to help prevent accidental burns. ‘Hot’ warning light which flashes whenever the plate temperature is above 50 °C, and operates even when the hotplate is turned off.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty