Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cole-Parmer® Incubators, Natural Convection, Antylia Scientific

Supplier: Antylia Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

52411-60 52411-58 52411-56 52411-64 52411-62
76539-142EA 1321.14 USD InStock
76539-142 76539-150 76539-148 76539-144 76539-146
Cole-Parmer® Incubators, Natural Convection, Antylia Scientific
Incubators Standard Incubators
Gravity convection provides a lower chance of sample cross contamination.

  • PID technology for high precision control and small temperature fluctuation
  • Extended-life silicone rubber gasket provides a long service life
  • Features corrosion-resistant stainless steel
  • Anti-skid shelf design, easy to operate by single hand
  • Glass doors ensure perfect observation of the samples

Gravity convection incubators allow heat to fill the chamber naturally, providing a lower chance of cross contamination between samples. These gravity convection incubators have a temperature range from ambient +5 to 65 ℃. The inner chamber is constructed of stainless steel with a round, curved inner angle for easy cleaning. The anti-skid shelf design is easy to use with a single hand. Smaller models have a large outer glass door while larger models have an internal glass door. Glass doors avoid unnecessary heat dissipation and temperature changes when samples need to be viewed. The LCD display shows time, set temperature, actual temperature, and running status. Safety features include an auto start feature after power loss/return, temperature deviation alarm, over-temperature set point alarm, and over current protection.

Temperature resolution: 0.1 °C

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty