Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cole-Parmer® Incubators, Forced Convection, Antylia Scientific

Supplier: Antylia Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

52411-70 52411-66 52411-72 52411-68 52411-76 52411-74 52411-78
76539-152EA 1279.05 USD InStock
76539-152 76539-162 76539-154 76539-164 76539-160 76539-156 76539-158
Cole-Parmer® Incubators, Forced Convection, Antylia Scientific
Incubators Standard Incubators
Mechanical convection circulates heat to all levels for more efficient heat distribution.

  • PID technology for high precision control and small temperature fluctuation
  • Extended-life silicone rubber gasket provides a long service life
  • Stainless steel inner chambers resists corrosion and is easy to clean
  • Anti-skid shelf design is easy to operate with a single hand
  • Glass doors ensure perfect observation of the samples

Mechanical convection offers faster heat up and better temperature uniformity and recovery due to the constant circulation of forced air. These mechanical convection incubators have a temperature range from ambient +5 to 65 ℃. The inner chamber is constructed of stainless steel with a round, curved inner angle for quick cleaning. The anti-skid shelf design is easy to use with a single hand. Smaller models have a large outer glass door while larger models have an internal glass door. Glass doors avoid unnecessary heat dissipation and temperature changes when samples need to be viewed.

Easy-to-view LCD shows time, set temperature, actual temperature, running status, and fan status. Safety features include auto-start after power loss/return, temperature deviation alarm, over-temperature set point alarm, and overcurrent protection.

Temperature resolution: 0.1 °C

Timer range: 0 to 5999 min

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty