Would you like to log in again?
Stainless Steel Dewar Flasks, Crystal Industries
Supplier: Crystal
SDF-1000 SDF-500
76539-166EA 133.01 USD InStock
76539-166 76539-168
Stainless Steel Dewar Flasks, Crystal Industries
Dewars and Cryogenic Vessels
These Stainless Steel Dewar Flasks are ideal for the storage and transport of low-temperature liquids and gases, or high-temperature liquids.
  • Temperature range: −190 to 200 °C
  • Available in 2 sizes (500 ml and 1000 ml)
  • Pharmaceutical grade stainless steel construction
  • Complete with vented stainless steels lids
  • Ideal for hot liquids, Liquid Nitrogen (LN₂), dry ice, CO₂ and other cryogenic gases
  • 2-year replacement warranty
Safely store and transport hot liquids, LN₂, dry ice, CO₂, or other cryogenic gases or allow small samples to cool.
The 1000 ml size container is equipped with a removable handle.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty