Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Cole-Parmer® SP Overhead Laboratory Mixers, Antylia Scientific

Supplier: Antylia Scientific
55008-02 55008-03
76539-174EA 3726.22 USD InStock
76539-174 76539-176
Cole-Parmer® SP Overhead Laboratory Mixers, Antylia Scientific
Stirrers Overhead Stirrers
Variable speed mixer designed for reliability and durability.

  • Durable - no wear user serviceable parts
  • Low/no maintanence brushless motor
  • Adjustable tilt from 0 to 30 degrees allows proper propeller placement
  • Hollow through shaft and user-replaceable keyless chuck
  • Adjustable mounting type

The Cole-Parmer SP laboratory mixer ensures accurate, dependable motor speed control with 'soft-start' feature and smooth, gradual adjustment to the speed setting. Constant speed is maintained electronically via automatic compensation for changes in load torque demands at both high and low speeds. Built-in motor current compensation minimizes speed drift with changes in load. Motor current limiting protects the system in case of a locked rotor condition. Output circuit protection protects against inadvertent output shorting.

The mixer rotates at 50 to 2500 rpm with a maximum torque of 91 in-oz. (64 Ncm) and has a through-shaft capability. Bracket hardware is preinstalled to enable installation of the adjustable tilt mounting bracket. The motor cable connector has a locking mechanism when the cable is connected to prevent unintended disconnection in the event of accidental force on the motor cable.

The variable-speed controller is used to turn on and set the speed and direction of the mixer motor. Easily switch the direction of rotation using the Forward-Off-Reverse switch. The LED indicates the mixer is powered by turning green and flashes when a limit condition has been reached.

Chuck: Keyless three-jawed type chuck
Controller dimensions (W×D×H): 9.7×9.9×3.5"

Motor type: Brushed DC motor
Motor dimensions (L×H): 3.4×10.9"

Certifications: IP 44 rated per IEC 60529.

Accessories information: 76539-174: Mixer motor (76539-186), bracket, mixer controller (76539-182), and power cord.
76539-176: Mixer motor (76539-186), bracket, mixer controller (76539-182), shaft ³/₈×18" (04552-35), 3" propeller (04552-60), double clamp (08041-22), stand (04552-80) and power cord.

Order Now
 

ORDER

ACCESSORIES

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty