Would you like to log in again?
Mag-Bind® PX blood RNA 96 kit, Omega Bio-Tek
Supplier: Omega Bio-Tek
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Mag-Bind®
M7763-00 M7763-00 M7763-01
76539-258EA 3056.57 USD InStock
CA76539-256 76539-258 76539-256
Mag-Bind® PX blood RNA 96 kit, Omega Bio-Tek
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The Mag-Bind® PX blood RNA kit allows for the isolation of total RNA from 2,5 ml of whole blood that has been stored in PAXgene® Blood RNA tubes. Stabilised blood samples are first spun down and the nucleic acid pellets are washed and recollected. After washing, the nucleic acid pellet is dissolved into a specially formulated resuspension buffer. At this point the samples can be transferred to 96-well plates for processing in robotic liquid handlers and magnetic processors. High quality intact RNA can be isolated in less than 90 minutes.
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty