Would you like to log in again?
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Supplier: Qorpak
CAP-00078 272496 272626 272487 CAP-00066 272635 272494 CAP-00062 272630 CAP-00060 272627 272633 CAP-00067 CAP-00068 CAP-00073 CAP-00072 272634 CAP-00063 CAP-00069 272628 272495 272491 CAP-00061 CAP-00079 272629 272492 272493 CAP-00071 272632 CAP-00065 CAP-00074 272490 272631 272497 CAP-00075
75993-610CS 930.46 USD InStock
75993-610 76539-294 75993-612 16161-428 76539-296 76539-290 76539-292 75993-590 16161-360 76175-390 75993-606 76539-302 76313-928 76539-304 16161-541 75993-608 76539-288 75993-625 75993-602 76539-300 75993-604 76539-284 16161-519 75993-620 76539-286 75993-600 75993-622 76539-280 16161-337 76539-282 75993-618 16161-574 75993-614 76539-298 75993-616
Green Thermoset Screw Caps, F217 and PTFE Liner, Qorpak®
Closures Bottle Closures
Green Thermoset Plastic Caps with F217 and PTFE liners are known for providing the widest range of chemical compatibility.
  • The color green easily identifies the F217 & PTFE liner system
  • F217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
The F217 features a low-density foam core between two LDPE discs. They are applied to the back of a PTFE liner.
Some uses for these F217/PTFE lined caps include analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty