Would you like to log in again?
Black Phenolic Screw Caps, F217 and Fluoropolymer Resin Liner, Qorpak®
Supplier: Qorpak
CAP-00145 CAP-00191 CAP-00171 CAP-06690 CAP-00205 CAP-07459 CAP-00310 CAP-00222 283051 CAP-00276
16151-899CS 2075.01 USD InStock
16151-899 76500-058 76209-746 71001-586 76539-328 76539-314 76539-316 76539-310 76539-320 76539-312
Black Phenolic Screw Caps, F217 and Fluoropolymer Resin Liner, Qorpak®
Closures Bottle Closures
F217 and PTFE liners resist attack from virtually all chemicals at room temperature
The F217 features a low density foam core between two LDPE discs and are applied to the back of a PTFE liner. This provides a tight seal with a low moisture transmission rate. Analytical lab samples, high purity chemicals, strong acids, solvents, environmental samples, pharmaceuticals, and diagnostic reagents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty