Would you like to log in again?
Fab Anti-MS4A1 Recombinant Antibody [clone: Abz522]
Supplier: Rockland Immunochemicals
400-001-MT2 400-001-MT2
76539-394EA 533 USD InStock
76539-394 CA76539-394
Fab Anti-MS4A1 Recombinant Antibody [clone: Abz522]
Antibodies
Recombinant Anti-CD20 Fab fragment Antibody has been tested for use in Flow Cytometry. This antibody recognizes structured CD20 and will work in western blot when the protein has not been denatured with DTT or bMe. Although not tested, this antibody could be useful in in IHC and in in-vivo and other cellular assays. Specific conditions for reactivity should be optimized by the end user.
  • Expressed in E. coli cells
  • >95% pure by SDS-PAGE analysis
  • Tested for use in Flow Cytometry
CD20 is a 33-37 kD, four transmembrane spanning protein, also known as B1 and Bp35. CD20 is expressed on pre-B-cells, resting and activated B cells (not plasma cells), some follicular dendritic cells, and at low levels on a T cell subset. CD20 is heavily phosphorylated on activated B cells and malignant B cells. Homo-oligomeric complexes of CD20 are thought to form Ca2+ conductive ion channels in the plasma membrane of B cells. The CD20 molecule is involved in B-cell activation and is associated with various Src family kinases (Lyn, Lck, Fyn). It exists in a complex with MHC class I and II, CD53, CD81, and CD82. Anti-CD20 antibody, Rituximab, sold under the brand name Rituxan, is a medication used to treat certain autoimmune diseases and types of cancer. It is used for non-Hodgkin lymphoma, chronic lymphocytic leukemia (in non-geriatric patients), rheumatoid arthritis, granulomatosis with polyangiitis, idiopathic thrombocytopenic purpura, pemphigus vulgaris, myasthenia gravis and Epstein–Barr virus-positive mucocutaneous ulcers. Biosimilars of Rituxan include Blitzima, Riabni, Ritemvia, Rituenza (F.K.A. Tuxella), Rixathon, Ruxience, and Truxima. This antibody is made from a Rituximab Fab fragment making it specific, efficient, and effective. Recombinant Anti-CD20 Fab fragment Antibody is useful for researchers interested in autoimmune disease and cancer research.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty