Would you like to log in again?
Anti-VEGF-A Recombinant Antibody [clone: Abz5]
Supplier: Rockland Immunochemicals
400-001-MT5 400-001-MT5
76539-400EA 533 USD InStock
CA76539-400 76539-400
Anti-VEGF-A Recombinant Antibody [clone: Abz5]
Antibodies
Humanized Recombinant Anti-VEGFA ScFV fragment Antibody has been tested for use in Flow Cytometry, Western Blot, and ELISA. This antibody recognizes structured VEGF-A and will work in western blot when the protein has not been denatured with DTT or bMe. Although not tested, this antibody could be useful in in IHC and in in-vivo and other cellular assays. Specific conditions for reactivity should be optimized by the end user
  • Expressed in CHO cells
  • >90% pure by SDS-PAGE analysis
  • Tested for use in Flow Cytometry, Western Blot, and ELISA
VEGF is a potent mitogen in embryonic and somatic angiogenesis with specificity for vascular endothelial cells. VEGF forms homodimers and exists in four different isoforms. Overall, the VEGF monomer resembles that of PDGF, but its N-terminal segment is helical rather than extended. VEGF shares homologies of about 21% and 24% respectively with the A and B chains of human platelet-derived growth factor (PDGF), and has complete conservation of the eight cysteine residues found in both mature PDGF chains. The cysteine knot motif is a common feature of this domain. The homology is not reflected in function, however, since the cell types responsive to VEGF are distinct from those responsive to homo- and heterodimers of the PDGF chains. This protein is a glycosylated mitogen that acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. VEGF-A also has been shown to have effects on a number of other cell types (e.g. stimulation of monocyte/macrophage migration, neurons, cancer cells, kidney epithelial cells). VEGF-A is also a vasodilator; it increases microvascular permeability, and was originally referred to as vascular permeability factor. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. Antibody against VEGF-A (Bevacizumab or brand name Avastin) is approved for treatment of cancer by inhibiting angiogenesis. Ranibizumab, a Fab fragment of Avastin or so-called Lucentis is approved to treat the "wet" type of age-related macular degeneration (AMD, also ARMD), diabetic retinopathy, and macular edema due to branch retinal vein occlusion or central retinal vein occlusion. This antibody has its amino acid sequence like one of Brolucizumab (sold under trade names Beovu or Vsiqq) - a humanized single-chain antibody fragment for the treatment of neovascular (wet) age-related macular degeneration (AMD).
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty