Would you like to log in again?
Anti-PCP Mouse Balb/c Monoclonal Antibody [clone: 3F6]
Supplier: Rockland Immunochemicals
MOB091-01 MOB091-01
76539-410EA 528 USD InStock
CA76539-410 76539-410
Anti-PCP Mouse Balb/c Monoclonal Antibody [clone: 3F6]
Antibodies
Anti-Pneumocystis Carinii (MOUSE) Monoclonal Antibody was tested in IHC-P with positive control infected lung tissues. Cellular Localization: Alveolar spaces. The user is advised to validate the use of the products with their tissue specimens prepared and handled in accordance with their laboratory practices. Consult references (Kiernan, 1981: Sheehan & Hrapchak, 1980) for further details on specimen preparation.
  • Specific against an 82 kDa parasite specific polypeptide
  • Tested in IHC-P with positive control infected lung tissues
  • Reacts with P. carinii in the fixed human tissues
  • Cellular Localization: Alveolar spaces
Pneumocystis jiroveci pneumonia is an infection of the lungs. The disease used to be called Pneumocystis carini or PCP pneumonia. The infection typically affects immunocompromised systems. It is an extracellular fungal infection. Transmission seems to be through inhalation.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty