Would you like to log in again?
Anti-EBV Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Supplier: Rockland Immunochemicals
MOB194-01 MOB194-01
76539-412EA 528 USD InStock
76539-412 CA76539-412
Anti-EBV Mouse Monoclonal Antibody [clone: CS1, CS2, CS3, CS4]
Antibodies
Anti-LMP-1 (Epstein-Barr Virus) (MOUSE) Monoclonal Antibody was tested in IHC-P with positive control infected lymphoblastoma, Hodgkin’s lymphoma tissues. Cellular Localization: Cytoplasmic. The user is advised to validate the use of the products with their tissue specimens prepared and handled in accordance with their laboratory practices. This antibody stains strongly with EBV positive lymphoblastoid cell lines and EBV infected B cell immunoblasts in infectious mononucleosis. Consult references (Kiernan, 1981: Sheehan & Hrapchak, 1980) for further details on specimen preparation.
  • Specific to 60 kDa latent membrane protein (LMP-1)
  • Tested in IHC-P with positive control infected lymphoblastoma, Hodgkin’s lymphoma tissues
  • Cellular Localization: Cytoplasmic
Latent membrane protein 1 acts as a CD40 functional homolog to prevent apoptosis of infected B-lymphocytes and drive their proliferation. It functions as a constitutively active tumor necrosis factor receptor that induces the activation of several signaling pathways, including those of the NF-kappa-B family. LMP1 signaling leads to up-regulation of antiapoptotic proteins and provide growth signals in latently infected cells. It interacts with host UBE2I and subsequently affects the sumoylation state of several cellular proteins. For example, LMP1 induces the sumoylation of host IRF7 thereby limiting its transcriptional activity and modulating the activation of innate immune responses.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty