Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Gorilla® Gorilla Tape
Supplier: Essendant
GOR105629 GOR6100109
76540-082RL 5.29 USD InStock
76540-082 76540-080
Gorilla® Gorilla Tape
Tapes
Gorilla tape is three times stronger for a hold that lasts. Made with double-thick adhesive, strong reinforced backing, and a tough all-weather shell, this duct tape is great for projects and repairs both indoors and out.
  • Double-thick adhesive grips smooth, rough, and uneven surfaces
  • Natural rubber adhesive
  • Reinforced polyethylene-coated cloth backing has a tight weave but can still be torn by hand
  • Rugged, weather-resistant shell withstands moisture, UV rays, and temperature changes
Gorilla tape sticks to smooth, rough, and uneven surfaces, including wood, stone, stucco, brick, metal, and vinyl. Dispenser not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty