Would you like to log in again?
Sequential Sterilization Wraps, Halyard
Supplier: Halyard
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
68272 68040 10712 68224 68012 10818 10724 68030 68012 68240 10848 10824 68248 68145 68040 10824 10848 68148 68148 10836 68048 68145 10712 68236 10818 68024 68272 68224 10836 68030 68240 68048 10724 68024 68248 68236
76540-240CS 149.44 USD InStock
76540-240 76540-244 76540-222 CA76540-230 76540-220 76540-242 76540-226 76540-248 CA76540-234 76540-224 76540-246 CA76540-232 CA76540-238 CA76540-216 76540-228 CA76540-236 CA76540-214 CA76540-220 CA76540-242 76540-232 CA76540-240 76540-230 CA76540-224 CA76540-246 76540-214 76540-236 CA76540-222 CA76540-244 76540-234 CA76540-228 76540-218 CA76540-248 CA76540-226 76540-216 76540-238 CA76540-218
Sequential Sterilization Wraps, Halyard
Sterilization Wraps
Wrap made of patented low-lint SMS sterilization fabric that is tear-, flame-, lint-, and abrasion-resistant. Recommended for light linen packs, linen gowns, towel packs, single basins, and other light wrapping.
  • Industry standard for sterile processing teams around the world
  • POWERGUARD* technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Class 1 flammability rating
Since 1978, the HALYARD* Sterilization Wrap, made of strong reliable SMS nonwoven fabric, has been helping keep sterilized instruments and supply sets ready for use at a moment’s notice. The company pioneered the development of SMS fabric and today is one of the largest global suppliers. It’s good to know that individual medical and dental practices can also take advantage of the same performance demanded by major medical and surgery centers. All HALYARD* products are made in the USA.
Today, Halyard Health offers six grades of wrap fabric, from H100 to H500, to offer the precise protection required for specific needs. Choosing is easy, the higher the number, the stronger the wrap. HALYARD* is also available in smaller sizes that meet the needs of smaller dental or medical offices.
Certifications: Maintenance of package integrity (MPI) to meet FDA requirements. Instructions for Use to meet FDA guidelines.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty