Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

PUREZERO* HG3 Light Blue Sterile Nitrile Gloves, Halyard

Supplier: Halyard
CLN923270 CLN923285 CLN923210 CLN923285 CLN923290 CLN923260 CLN923270 CLN923260 CLN923275 CLN923265 CLN923210 CLN923265 CLN923275 CLN923280 CLN923290 CLN923280
76540-310CS 493.05 USD InStock
76540-310 CA76540-296 76540-298 76540-296 76540-302 CA76540-300 CA76540-310 CA76540-298 76540-300 CA76540-304 76540-306 CA76540-302 76540-304 CA76540-308 76540-308 CA76540-306
PUREZERO* HG3 Light Blue Sterile Nitrile Gloves, Halyard
Gloves
PUREZERO* Cleanroom Gloves are ideal for applications that involve handling delicate equipment in microelectronics, semiconductors, optics, pharmaceutical, and medical device manufacturing applications. They are specifically designed to meet the stringent requirements of cleanroom environments.

  • Recommended for use in ISO Class 3 or higher and Grade A/B/C/D cleanrooms
  • Maximum particles: >0.5 µm/cm² less than 1200
  • AQL 0.65 for pinholes
  • Accelerator-, silicone-, latex-, and powder-free
  • Hand specific, 30.5 cm (12") length with beaded cuff
  • Fingertip thickness: 0.10 mm/4 mil
  • Palm thickness: 0.08 mm/3.1 mil
  • Static for dissipative use, SAL 10⁻⁶

These gloves are clean processed (washed repeatedly in deionized water) to ensure consistent control of low particles, extractables, and endotoxin levels. They are recommended for use in ISO Class 3 or higher cleanroom environments. They are individually pair packaged for aseptic donning (poly wallet and pouch) and have triple-layer cleanroom compatible packaging (poly pouch, inner bag, and case liner). PUREZERO* HG3 Sterile Light Blue Nitrile Cleanroom Gloves have a hand-specific shape, 4 mil fingertip thickness, and a smooth finish, with beaded cuff to aid in donning and help prevent roll down. With gamma irradiation to a Sterility Assurance Level (SAL) of 10¯⁶they are suitable for aseptic cleanroom applications.

Because PUREZERO* HG3 sterile cleanroom gloves are made with an accelerator-free nitrile polymer, there is a reduced risk of allergies and skin irritation associated with accelerator chemicals in other nitrile gloves.

Certifications: Certificates of Analysis (CoA), and Certificates of Irradiation (CoI) easily accessible online for every production lot. Declaration of Conformity (DoC) documents showing compliance to applicable regulations and standards are also available.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty