Would you like to log in again?
Universal Precautions Lab Coats, Halyard
Supplier: Halyard
10046-66 10045 10041 10047-66 10041-66 10045-66 10040 10049 10048 10048-66 10040-66 10046 10044 10047 10042 10043-66 10042-66 10049-66 10044-66 10043
76540-372CS 299.04 USD InStock
76540-372 CA76540-370 CA76540-380 76540-370 76540-380 CA76540-374 76540-376 76540-364 CA76540-362 CA76540-372 76540-374 76540-362 CA76540-378 CA76540-366 76540-368 CA76540-376 76540-378 CA76540-364 76540-366 CA76540-368
Universal Precautions Lab Coats, Halyard
Disposable Garments
Halyard Health Lab Coats all feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Traditional collar
  • Knit cuffs
  • Six snap front closure
  • Three reinforced pockets
  • Size/name label
Halyard Health Lab Coats all feature a back vent for added comfort during extended use, and a neck loop for hanging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty