Would you like to log in again?
Universal Precautions Lab Jackets
Supplier: Halyard
10071 10073 10072 10069
76540-382CS 229.13 USD InStock
76540-382 76540-386 76540-384 76540-388
Universal Precautions Lab Jackets
Disposable Garments
Halyard Health lab jackets feature a protective 3-layer fabric that offers exceptional resistance to blood strike-through at various levels of exposure.
  • Knit collar and cuffs
  • 5 snap front closure
  • 3 reinforced pockets
  • Raglan sleeves
  • Size/name label
Made from a protective 3-layer SMS fabric, all lab jackets feature a convenient neck loop for hanging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty