Would you like to log in again?
Tri-Layer AAMI2 Isolation Gowns, Halyard
Supplier: Halyard
69981 69988 69979
76540-394CS 234.98 USD InStock
76540-394 76540-398 76540-396
Tri-Layer AAMI2 Isolation Gowns, Halyard
Disposable Garments
These Isolation Gowns provide protection based on AAMI guidelines, the most widely accepted system of classification, determined by liquid barrier performance, for protective apparel and surgical drapes.
  • Available in blue or yellow
Three levels of protection allow users to select the right gown for the procedure at the right price.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty