Would you like to log in again?
CHEMO 360* Procedure Gowns, Halyard
Supplier: Halyard
47346 47347
76540-408CS 575.81 USD InStock
76540-408 76540-410
CHEMO 360* Procedure Gowns, Halyard
Disposable Garments
Gear up for the fight now because there is no safe level of exposure to cytotoxic chemotherapy drugs for healthcare workers.
  • Tested against 52 chemotherapy drugs on fabric and seams
  • Meets USP <800> guidelines and passes ASTM F739-12
  • Fabric passes ASTM F1670 & F1671, resistance against fluid and bloodborne pathogen penetration
  • Polyethylene coated spunbound fabric
  • Full back design with tape-tab neck closure
  • Knit cuffs
New, stricter guidelines on hazardous drugs handling in a healthcare setting, USP <800>, applies to all healthcare personnel with no exceptions. Choose the HALYARD* CHEMO 360* Chemotherapy Procedure Gown to protect against 52 chemotherapy drugs.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty