Would you like to log in again?
Laser Cassette Printers, VEGA/NOVA
Supplier: Epredia
8000039 8000038 NOVA05-FT-PCDI VEGA05-FT-PCDI
76540-418EA 62110.88 USD InStock
76540-418 76540-420 76540-446 76540-444
Laser Cassette Printers, VEGA/NOVA
Printers Cassette and Slide Printers
The Epredia Laser Cassette Printing technology produces high quality print combined with a patented chemical additive in the cassette to deliver consistency and efficiency. Built in interface connectivity ensures LIS/LIM compatibility.
  • Epredia NOVA laser Printer is a single cassette hopper printer providing an economical option for laboratories
  • Three additional hoppers are included for convenient cassette exchanges
  • Epredia VEGA laser printer has six cassette hoppers and supports laboratories printing a variety of cassette colors and styles
  • Laser Technology eliminates ancillary components such as print heads & ribbons and simplifies operation
  • High resolution printing, 600 dpi, improves printing sharpness and clarity of characters allowing more characters to be embedded on cassette surface
  • Fully automated system with fast print speed, five seconds per cassette, allows the user to quickly and easily print cassettes needed for a case at accessioning or grossing
  • Laser system with cassette additive print technology ensures permanent and delible marking that are extremely durable overtime
  • Small footprint reduces the concern of space constraint in the lab
  • Quiet printing reduces distraction during grossing and accessioning
Workflow Efficiencies –
Epredia laser printers are designed to create crisp, accurate and indelible print quality for laboratories concerned about workflow bottlenecks caused by the need for reprinting cassettes with low-quality or unreadable printing at accessioning.
High Printing Quality –
•Epredia laser printers create sustainable and indelible printed information where patient data can be identified accurately throughout the lifetime of the cassette supporting laboratories and patients.
•Character Types: Alphanumeric.
•Barcode Types: Linear, Bidimensional, ASCII Codes.
High Throughput –
Epredia laser printers incorporate a highly efficient technology helping labs with higher printing demands utilize the laser printing capabilities and automated cassette feeder feature to achieve high throughput and faster turn-around times.
Low Operations Cost –
Epredia laser printers do not require replacement of ancillary components significantly decreasing operation costs and lowers any intensive maintenance helping lab managers from being burdened with high operation costs.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty