Would you like to log in again?
NEBNext® Library Quant DNA Standards - 6 Standards, New England Biolabs
Supplier: New England Biolabs
NEBNext®
E7642S E7642S
76540-504EA 190 USD InStock
CA76540-504 76540-504
NEBNext® Library Quant DNA Standards - 6 Standards, New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
NEBNext Library Quant DNA Standards are a set of six standards, defining a curve from 100 pM to 0.001 pM, intended for use with the NEBNext Library Quant Master Mix found in the NEBNext Library Quant Kit for Illumina (NEB #E7630).
  • Additional volume of the standards included in NEB #E7630
  • qPCR-based library quant based on either four or six standards
The six standards offered are the same as the six that are included in the base kit. The NEBNext Library Quant Kit manual includes details for using either four or six standards for quantitation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty