Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid 36.5-38%, Low-Metal (LM) grade semiconductor grade

Supplier: Transene
Danger
Synonyms: Muriatic acid

HCL0000-GAL HCL0000-QT
76541-062EA 21.29 USD InStock
76541-062 76541-028
Hydrochloric acid 36.5-38%, Low-Metal (LM) grade semiconductor grade
Hydrochloric acid
Transene Hydrochloric Acid LM is a "low metals" HCl solution suitable for demanding semiconductor and electronics processing.


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 109 °C
Melting Pt: -74 °C
Density: 1.16 g/cc
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay (HCl) 36.5-38.0 %
Ammonium (NH4) 500 ppb max.
Color 7 APHA max.
Phosphate (PO4) 50 ppb max.
Free Chloride (Cl) 500 ppb max.
Heavy Metals 100 ppb max.
Bromide (Br) 50000 ppb max.
Extractable Organics 500 ppb max.
Residue After Ignition 3000 ppb max.
Aluminum (Al) 10 ppb max.
Arsenic and Antimony (as As) 5 ppb max.
Barium (Ba) 10 ppb max.
Beryllium (Be) 10 ppb max.
Bismuth (Bi) 10 ppb max.
Boron (B) 10 ppb max.
Calcium (Ca) 5 ppb max.
Chromium (Cr) 10 ppb max.
Cobalt (Co) 10 ppb max.
Copper (Cu) 10 ppb max.
Gallium (Ga) 10 ppb max.
Germanium (Ge) 10 ppb max.
Gold (Au) 10 ppb max.
Indium (In) 10 ppb max.
Iron (Fe) 10 ppb max.
Lead (Pb) 10 ppb max.
Lithium (Li) 10 ppb max.
Magnesium (Mg) 10 ppb max.
Manganese (Mn) 10 ppb max.
Molybdenum (Mo) 10 ppb max.
Nickel (Ni) 10 ppb max.
Platinum (Pt) 10 ppb max.
Potassium (K) 10 ppb max.
Silver (Ag) 10 ppb max.
Sodium (Na) 10 ppb max.
Strontium (Sr) 10 ppb max.
Sulfate (SO4) 500 ppb max.
Sulfite (SO3) 800 ppb max.
Titanium (Ti) 10 ppb max.
Zinc (Zn) 10 ppb max.
Zirconium (Zr) 10 ppb max.
Particle Count, 0.5 mm 50 ct/ml max.
Particle Count, 1.0 mm 10 ct/ml max.

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty