Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CryoCube® F440 ULT Freezers, 115 V

Supplier: Eppendorf
CryoCube®
F440202035 F440243035 F440243035 F440203035 F440200035 F440240035 F440202035 F440242035 F440240035 F440242035 F440200035 F440203035
76542-106EA 18991.63 USD InStock
76542-106 CA76542-104 CA76542-114 76542-104 76542-114 CA76542-112 76542-112 CA76542-110 76542-110 CA76542-108 CA76542-106 76542-108
CryoCube® F440 ULT Freezers, 115 V
Freezers Ultra Low Temperature Freezers
The Eppendorf CryoCube F440 ULT freezers offer a combination of minimal operating costs and superior sample protection. With a five-compartment layout and five inner doors for easy sample access, the solid insulation shields your valuable samples.

  • 115 V model
  • Volume: 440 L (15.5 cu. ft.), e.g. up to 32000 samples
  • Efficient insulation, composed of traditional insulation ('n'-version) and vacuum insulation panels ('h'-version), saves energy
  • Automatic vent port on front door of upright CryoCube models improves energy consumption and enables fast sample access
  • Lockable main switch to prevent switch-offs by accident
  • Location for chart recorder for data documentation
  • Stainless steel interior for easy cleaning during maintenance and defrost cycles; powder-coated steel exterior

Ergonomic door handle of CryoCube models based on Eppendorf PhysioCare Concept® requires less force and provides easier access. Efficient compressor control system reduces cycle times to lower energy consumption and increases freezer longevity.

The Eppendorf CryoCube F440 series offers a five compartment lay-out and five insulated inner doors for easy sample access and safe sample storage at −80 °C.

The ULT freezer has a temperature range from −50 to −86 °C. The CryoCube F440 series is based on two different freezer models:
CryoCube F440n:
• 130 mm wall (polyurethane insulation)
• Air-cooling
CryoCube F440h:
• 130 mm wall (polyurethane insulation and vacuum insulation panels)
• Air-cooling

Heat output:
F440h: 271 W
F440n: 354 W

Max. power consumption:
F440h: 6500 W
F440n: 8500 W

Pull-down time to −80 °C: 3 h

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty