Would you like to log in again?
Digital Dry Baths, Crystal
Supplier: Crystal
DB-04A DB-03A DB-02A DB-01A
76542-744EA 590.87 USD InStock
76542-744 76542-742 76542-748 76542-746
Digital Dry Baths, Crystal
Block Heaters Digital Block Heaters
Dry baths are versitale, reliable, and designed for precise temperature control for processing precious samples in a variety of applications with 1-, 2- and 4-block models.
  • Digital display of temperature
  • Precise temperature control
  • Fast heat-up times
  • Programmable models DB-01A, DB-03A and DB-04A offer 6 segments with timer
  • Overheating alarm and automatic shutoff on all models for added safety
  • Large selection of blocks for tubes, vials and microplates available
  • Includes cover(s) for improved temperature uniformity
All Crystal Dry Bath Heaters offer the confidence and effeciency that your molecular biology, histology, clinical, environment and industrial applications demand. See the list of blocks available.
Two-year warranty.
Certifications: CE, RoHS, WEEE.
Ordering information: Blocks sold separately.
Delivery information: Supplied with models DB-01A and DB-02A: 1× DBA05, model DB-03A: 2× DBA05 and model DB-04A: 4× DBA05 insulating cover.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty