Would you like to log in again?
Rapid Response™ FIT Mailers, BTNX
Supplier: BTNX
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Rapid Response™
FOB-0M
76543-130CT 1214.46 USD InStock
76543-130
Rapid Response™ FIT Mailers, BTNX
Clinical Diagnostic Supplies Clinical Reagents and Kits
The Rapid Response™ fecal immunochemical test (FIT) mailer is a paper kit designed to be given to patients for at-home collection of spontaneously passed stool specimens (SPS). This easy collection method increases patient compliance and ensures an appropriate sample.
  • Enables easy at-home collection
  • Ensures appropriate sample collection
Use in conjunction with the Rapid Response™ FIT test (VWR Cat. No. 76473-472) and the Rapid Response™ IFOBT external controls (VWR Cat. No. 76473-474).
Ordering information: Kit does not include Rapid Response™ FIT test or the Rapid Response™ IFOBT external controls.
Packaging: 50 mailers.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty