Would you like to log in again?
PROSAT® Meltblown Polypropylene Wipes with CyQuanol™, Contec®
Supplier: Contec
PROSAT®
PSPPCQ01 PSPPCQ02
76543-176CS 1226.9 USD InStock
76543-176 76543-178
PROSAT® Meltblown Polypropylene Wipes with CyQuanol™, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
Presaturated with quaternary ammonium and ethyl alcohol.
  • Meltblown polypropylene fabric provides exceptionally clean and free from additives
  • 1 minute disinfectant contact time is easy to maintain compliance and appropriate dwell time
  • Low in particles and fibers makes it ideal for a variety of critical applications
  • Presaturated with disinfectant reduces VOC emissions and solvent use
  • Resealable pouch preserves cleanliness and solvent saturation levels
PROSAT® Wipes with CyQuanol™ are intended for Life Science cleanroom manufacturers who want to effectively maintain cGMP compliance by maximizing efficiency and minimizing inconsistency in their contamination control process.
PROSAT® Meltblown Polypropylene Wipes with CyQuanol™ provide a consistent release of solvent to thoroughly disinfect surfaces in critical environments. They are exceptionally clean and also free from additives of any kind. Meltblown polypropylene contains very low levels of sodium and other ions. These wipes are EPA registered with proven efficacy claims against bacteria, viruses, and fungi. With a short contact time of 1 minute, these wipes make compliance to disinfection protocols more manageable, as microbial kill claims can be achieved without reapplication. Using presaturated wipes delivers a controlled amount of disinfectant to surfaces every time compared to traditional spray methods. Presaturated wipes ensure repeatability and reproducibility of disinfectant protocols to improve process consistency.
PROSAT® Wipes with CyQuanol™ are also available with polyynic heat-seal. Both substrates are available sterile for the most critical areas.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty