Would you like to log in again?
IV Stands, Blickman
Supplier: Blickman
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
0561350400 0539A02302 0561370400 0537792004 0517794400 0561305401 0517794000 0571410000 0561305000 0541371600 0561350200 0561310000 0531415400 0541370400 0541370600 05413710RH 0531415000 0561315400 0561370600 0537792002 0518889400 0539A023RH 0561350600 0571410400 0541110100 0537792000 0539A02304 0541110200 0528890000 0528890400 05413700RH 0537792400 0518889000 0561370RH1 0541371400 0561315000
76545-262EA 399.91 USD InStock
76545-262 76545-284 76545-286 76545-264 76545-300 76545-266 76545-288 76545-302 76545-268 76545-280 76545-260 76545-282 76545-314 76545-316 76545-318 76545-296 76545-274 76545-252 76545-298 76545-254 76545-276 76545-278 76545-256 76545-310 76545-258 76545-312 76545-290 76545-292 76545-270 76545-272 76545-250 76545-294 76545-304 76545-248 76545-306 76545-308
IV Stands, Blickman
Support Stands
IV poles are available in high quality chrome and quality 304 stainless steel, and set on 4, 5 or 6 leg bases with casters, no projections and great balance/tip resistance.
  • Bases designed to spin 360˚ in tight spaces with great balance
  • Some bases are gravity weighted to help compensate for heavier loads
  • 2, 4, 6, or 8 (rake) hooks available for most IV pole models
  • All models come with colored secure grip tips
Our IV poles are optimal for maintaining continuous fluid delivery for every patient, available in a wide range of adjustable heights with a variety of easy locking options that can be operated with one hand.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty