Would you like to log in again?
PMSKN Parallel Artificial Membrane Permeability Assay-Skin Kit, BioAssay Systems
Supplier: BioAssay Systems
PMSKN-096
76545-504KT 455.38 USD InStock
76545-504
PMSKN Parallel Artificial Membrane Permeability Assay-Skin Kit, BioAssay Systems
Assays
Assess skin membrane permeability of test compounds.
  • Convenient: Includes all necessary equipment to run a PAMPA plate
  • Simple and low-cost: Procedure is easy to follow and more affordable than cell-based permeability assays
  • High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: PAMPA
  • Samples: Chemical compounds
MEMBRANE PERMEABILITY is an important characteristic to determine for evaluating compounds as potential drug candidates. Drugs often need to cross cell membranes in order to reach their target of action and this makes a compound’s ability to passively cross these membranes an important characteristic to evaluate. The skin, and in particular, the stratum corneum is a complex barrier, which can be mimicked.
Rapid and early screening of compounds for skin penetration is highly desirable for drug discovery. Permeability can be evaluated by cell-based methods; however, these methods are often expensive and time consuming. Parallel Artificial Permeability Assays (PAMPA) offer researchers a quick, inexpensive method of evaluating the permeability of test compounds. Our PMSKN-096 kit is designed to aid in evaluating skin permeability.
BioAssay Systems PMSKN Kit provides all the necessary components to run a Parallel Artificial Permeability Assay for skin permeability studies.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty