Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
EnzyFluo™ NADP+/NADPH Assay Kit, BioAssay Systems
Supplier: BioAssay Systems
E2NP-100
76545-526KT 517.1 USD InStock
76545-526
EnzyFluo™ NADP+/NADPH Assay Kit, BioAssay Systems
Assays Nucleic Acid Assays
Direct Assays: NADP+/NADPH concentrations and ratios in cell or tissue extracts.
  • Sensitive and accurate: Detection limit 0.1 µM, linearity up to 10 µM NADP+/NADPH in 96-well plate assay
  • Convenient: The procedure involves adding a single working reagent, and reading the optical density at time zero and 30 min at room temperature
  • High-throughput: Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day
  • Method: Colorimetric Determination at 565 nm
  • Samples: Cells, Tissue
  • Species: human, mouse, rat and more
Pyridine nucleotides play an important role in metabolism and, thus, there is continual interest in monitoring their concentration levels. Quantitative determination of NADP+/NADPH has applications in research pertaining to energy transformation and redox state of cells or tissue.
Simple, direct and automation-ready procedures for measuring NADP+/NADPH concentration are very desirable. BioAssay Systems' EnzyChromTM NADP+ /NADPH assay kit is based on a glucose dehydrogenase cycling reaction, in which the formed NADPH reduces a formazan (MTT) reagent. The intensity of the reduced product color, measured at 565 nm, is proportionate to the NADP+/NADPH concentration in the sample. This assay is highly specific for NADP+/NADPH and is not interfered by NAD+/NADH. Our assay is a convenient method to measure NADP, NADPH and their ratio.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty