Would you like to log in again?
Super Erecta Lab Utility Carts, Stainless Steel Solid Shelves, Metro
Supplier: Metro International
MW108L MW208L MW204L MW104L
76545-660EA 2950.68 USD InStock
76545-660 76545-662 76545-664 76545-666
Super Erecta Lab Utility Carts, Stainless Steel Solid Shelves, Metro
Carts
Durable materials and total-guard polymer/stainless steel casters for long life in laboratory environments.
  • Total Guard all polymer and stainless-steel casters 2 swivel/2 total lock
  • All type 304 stainless steel shelves with epoxy-coated corners
  • Two stainless steel handles
Features Super Erecta type 304 stainless steel solid shelves with epoxy-coated corners and two stainless handles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty