Would you like to log in again?
Racks for Freezers
Supplier: Eppendorf
6001062910 6001011510 6001081110 6001061910 6001071510 6001072510 6001000310 6001000410 6001050110 6001021110 6001050211 6001011910 6001040210 6001021110 6001022910 6001012210 6001081310 6001000911 6001000410 6001012310 6001021210 6001050110 6001072410 6001000910 6001071410 6001021910 6001000510 6001081510 6001000910 6001072210 6001061310 6001012410 6001081410 6001071310 6001022310 6001082910 6001021410 6001000310 6001000510 6001050011 6001040011 6001081310 6001040211 6001021310 6001082510 6001012310 6001050211 6001072910 6001062510 6001061510 6001071210 6001050210 6001000411 6001022410 6001050210 6001021310 6001012210 6001081210 6001072510 6001012510 6001011210 6001050111 6001021210 6001072310 6001022210 6001071210 6001011310 6001071110 6001022410 6001011410 6001000311 6001012910 6001072910 6001000911 6001040010 6001062210 6001081910 6001021410 6001082210 6001022910 6001082910 6001071310 6001072410 6001011210 6001081510 6001012510 6001071510 6001011510 6001040111 6001011910 6001071910 6001082310 6001082510 6001022310 6001011110 6001071910 6001011110 6001081910 6001062410 6001040110 6001062310 6001061410 6001000311 6001072310 6001011310 6001021910 6001071410 6001072210 6001000411 6001071110 6001081210 6001012410 6001050011 6001050111 6001011410 6001081410 6001061210 6001082410 6001082410 6001081110 6001050010 6001050010 6001022210 6001082210 6001012910 6001082310
76545-774EA 323.85 USD InStock
CA76545-772 CA76545-770 CA76545-806 CA76545-804 CA76545-808 CA76545-766 CA76545-764 CA76545-802 CA76545-800 CA76545-768 CA76545-780 CA76545-784 CA76545-782 76545-774 76545-806 76545-808 CA76545-816 CA76545-776 CA76545-810 CA76545-774 76545-800 CA76545-814 76545-802 CA76545-778 76545-804 CA76545-812 76545-780 CA76545-790 76545-782 CA76545-794 76545-784 CA76545-792 76545-786 CA76545-708 CA76545-706 76545-810 CA76545-700 CA76545-788 76545-776 76545-812 76545-778 CA76545-786 76545-814 CA76545-704 76545-816 CA76545-702 CA76545-680 76545-790 76545-792 CA76545-684 76545-794 76545-676 CA76545-682 76545-796 CA76545-718 76545-708 CA76545-716 76545-700 76545-788 CA76545-798 CA76545-710 CA76545-678 CA76545-796 76545-702 CA76545-676 CA76545-714 76545-704 CA76545-712 76545-706 76545-680 CA76545-692 76545-682 CA76545-690 76545-684 CA76545-696 76545-720 76545-686 CA76545-694 76545-718 CA76545-728 CA76545-688 76545-678 76545-798 CA76545-722 76545-710 CA76545-686 76545-712 CA76545-720 CA76545-726 76545-714 76545-716 CA76545-724 76545-692 76545-694 76545-696 CA76545-740 76545-698 76545-690 CA76545-738 CA76545-732 76545-722 76545-688 76545-724 CA76545-730 CA76545-698 76545-726 CA76545-736 76545-728 CA76545-734 CA76545-750 76545-742 CA76545-744 CA76545-742 CA76545-748 CA76545-746 CA76545-762 76545-750 76545-752 CA76545-760 76545-744 CA76545-754 76545-746 CA76545-752 76545-748 CA76545-758 CA76545-756
Racks for Freezers
Racks Cryogenic Box Racks
These racks are heavy-duty and designed for higher capacity and durability. Specially designed for optimised cooling conditions and improved sample safety.
  • Fit freezer boxes up to a 136×136 mm for increased storage flexibility
  • Drawer-style freezer racks, each drawer can be individually removed, leaving the rest in the freezer
  • In side-access racks each box fits into a distinct, individual compartment for easier inventory tracking
There are two systems used in laboratories - the standard 4-boxes deep racks and 5-boxes deep racks. The easiest way is to take out all racks (with the samples/ boxes included) and store those racks in some backup freezer. To be compatible with most common ULT freezer suppliers, these ULT freezers are designed for 4-boxes deep racks.
The ULT freezer with 3-compartments/shelves has higher compartments compared to the 5-compartment ULT freezer. Racks of a 5-compartment freezer do fit into a 3-compartment freezer but not vice versa. Most of the storage boxes have to be pulled out of the in the cold inner area of the ULT freezer to access the samples.
To get the maximum capacity out of the ULT freezer, the CryoCube F740 series (3-compartment) can be equipped with 3-compartment MAX racks. These racks only fit into the CryoCube F740 series 3-compartment instruments.
Delivery information: Locking rods are now available for chest freezer racks to provide additional sample security.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty