Would you like to log in again?
Vee Gee Glass Stoppers
Supplier: Heathrow Scientific
03083 03082 03087 03085 03081 03084
76545-850PK 61.14 USD InStock
76545-850 76545-840 76545-842 76545-844 76545-846 76545-848
Vee Gee Glass Stoppers
Stoppers Glass Stoppers
These stoppers are lightweight yet very strong to reduce chipping and breakage, thus minimizing replacement costs.
  • Meet ASTM E-675 specifications
  • Flat top design allows stopper to stand on its head
  • Fully interchangeable with other common stoppers
  • Clear color
  • Comes in packs of 6
The flat top design allows the stopper to stand on its head, which frees the user’s hand and minimizes potential contamination. They are fully interchangeable with other common flask stoppers and fit all standard labware of comparable size.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty