Would you like to log in again?
Handheld Sodium Chloride Refractometers
Supplier: Heathrow Scientific
43033 43035
76545-870EA 124.85 USD InStock
76545-870 76545-868
Handheld Sodium Chloride Refractometers
Refractometers Handheld Refractometers
The handheld refractometer is exceptionally simple to operate and provides quick, accurate measurements for the concentration of nearly any aqueous solution in a wide range of applications.
  • Large, sharp display scales with high contrast
  • High-quality optical glass material for consistent, accurate readings
  • Durable, industrial-grade design with non roll stand feature to keep unit secure on flat surfaces
  • Tactical, non-slip rubber grip on body to block transmission of hand heat reducing temperature effects
  • Prism encased in all-metal housing allowing sample and prism to reach temperature equilibrium quickly
  • Maximum sample temperature: 90 °C (194 °F)
  • 5 year warranty
The handheld sodium chloride refractometers feature the largest, easiest-to-read scale available with clear, sharp figures, screen shading and incredible contrast. This significantly enhances overall precision, even when several different people use the refractometer. These sodium chloride refractometers also come with a rubber grip on the body, which blocks the transmission of hand heat to eliminate temperature errors.
The rugged construction of these handheld sodium chloride refractometers will provide years of reliable use. The optical glass prism is mounted in an all-metal housing which allows the sample and prism to reach temperature equilibrium quickly. The non-roll stand ensures that once these sodium chloride refractometers are placed on a table or bench top, they won't be prone to roll off the surface.
Delivery information: Supplied with a zippered vinyl carrying case, plastic transfer pipette and instruction manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty