Would you like to log in again?
Digital Refractometer, DEF/Battery Acid, 3 Scales, MDX-602
Supplier: Heathrow Scientific
48602
76545-894EA 450.21 USD InStock
76545-894
Digital Refractometer, DEF/Battery Acid, 3 Scales, MDX-602
Refractometers Handheld Refractometers
This digital urea (DEF) and battery acid refractometer has 3 scales. It measures urea (DEF), refractive index, and battery acid. It's used to ensure compliance with remissions for diesel fuel powered vehicles.
  • Compact and lightweight
  • Removable grip and prism cover for easy cleaning
  • Continuous loop read features (15 reads)
  • Automatic temperature control
  • Zero with distilled water
  • 589.3 nm LED
  • Stainless steel prism well
  • °C/°F switchable temperature display
  • 1 year warranty
The MDX-Series Digital DEF Battery Acid Refractometer features IP 65/67 ratings making it ideal for use in the field and in a multitude of environments. Delivering quick, digital accuracy with high-end performance features such as Automatic Temperature Compensation (ATC) and Continuous Loop Average Readings.
This unit will also monitor the strength of battery acid and specific gravity. Common applications for this refractometer include the trucking industry, farm/agriculture equipment, construction equipment, and other vehicles that use diesel fuel.
Certifications: IP protection class according to DIN EN 60529: IP 67 (for detecting unit) and IP 65 for remainder
Delivery information: Supplied with digital refractometer (Model MDX-602), rubber grip and prism cover, hard shell case with protective lining, disposable pipet, battery and user manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty