Would you like to log in again?
Digital Refractometer, Glycols/Coolant, 4 Scales, MDX-603
Supplier: Heathrow Scientific
48603
76545-896EA 445.02 USD InStock
76545-896
Digital Refractometer, Glycols/Coolant, 4 Scales, MDX-603
Refractometers Handheld Refractometers
This digital coolant refractometer measures the concentration and freezing point of the coolants ethylene glycol and propylene glycol. This unit has 4 scales (ethylene glycol, ethylene glycol freeze point, propylene glycol, and propylene glycol freeze point) and comes with a 1-year warranty.
  • Compact and lightweight
  • Removable grip and prism cover for easy cleaning
  • Continuous loop read features (15 reads)
  • Automatic temperature control
  • Zero with distilled water
  • 589.3 nm LED
  • Stainless steel prism well
  • °C/°F switchable temperature display
  • 1 year warranty
The MDX-Series Digital Brix/Sugar Refractometer features IP 65/67 ratings making it ideal for use in the field and in a multitude of environments. Delivering quick, digital accuracy with high-end performance features such as Automatic Temperature Compensation (ATC) and Continuous Loop Average Readings.
Certifications: IP protection class according to DIN EN 60529: IP 67 (for detecting unit) and IP 65 for remainder.
Delivery information: Supplied with digital refractometer (Model MDX-603), rubber grip and prism cover, hard shell case with protective lining, disposable pipet, battery and user manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty