Would you like to log in again?
Serialized Glass Thermometers
Supplier: Heathrow Scientific
80905 80903 80703 80907 80708 80701 80910 80706 80906 80908 80902 80901 80702 80707
76545-906EA 27.73 USD InStock
76545-906 76545-916 76545-908 76545-918 76545-920 76545-898 76545-922 76545-900 76545-910 76545-902 76545-924 76545-912 76545-904 76545-914
Serialized Glass Thermometers
Thermometers Analog Thermometers
Our precision, serialized thermometers are ideal for use in laboratories, universities, food/beverage, environmental, wastewater, and many other general-purpose applications requiring serialization.
  • Feature safety blue fill against a white background for easy reading
  • Wide variety of temperature ranges available
The thermometer features a safety blue fill, which is an organic performance liquid that is bio-degradable and exhibits all the qualities of mercury without the risk of contamination and hazards associated with mercury.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty