Would you like to log in again?
Dial Thermometers, 1" Diameter
Supplier: Heathrow Scientific
81110 81125 81550 81070
76545-928EA 5.3 USD InStock
76545-928 76545-930 76545-932 76545-926
Dial Thermometers, 1" Diameter
Thermometers Analog Thermometers
Our dial thermometers are an excellent choice for many applications including food and beverage quality control, general lab use, and asphalt/concrete testing.
  • Small stem fits into cover (included) for safe attachment to a pocket or holder
  • Compact design for convenient use on the go
  • Durable polycarbonate lens withstands wear and tear
The small stem and face fits nicely into a shirt pocket or holder. A wide variety of temperature ranges are available. It comes with a stem cover for simplified transport and storage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty