Would you like to log in again?
Digital Stem Thermometers
Supplier: Heathrow Scientific
83210-12 83110
76545-942EA 24.32 USD InStock
76545-942 76546-706
Digital Stem Thermometers
Thermometers Digital Thermometers
Our digital stem thermometers are ideal for temperature monitoring in essentially any setting. Ideal for easily monitoring reagents and liquids in test tubes, beakers, and flasks, food processing, refrigeration, horticulture and much more.
  • The 83000-series features a dual scale (°C/°F), fast 1-second update time
  • Swiveling head can be moved a full 180° for easy reading in any position
  • Both the ON/OFF switch and the scale selection switch are located on the face for simple access
The thermistor sensor stems are stainless steel, 3.5 mm (0.138") diameter and come with a protective case featuring a pocket clip.
Delivery information: Includes a 1.5 V alkaline coin cell battery (Type LR44) and features automatic power-off after 10 minutes of idle use.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty