Would you like to log in again?
VanGuard 1300-Series, Compound Microscopes, Vee Gee Scientific
Supplier: Vee Gee Scientific
Vanguard
1323PHI 1331BRI 1333PHI 1321BRI
76545-990EA 1749.73 USD InStock
76545-990 76545-992 76545-986 76545-988
VanGuard 1300-Series, Compound Microscopes, Vee Gee Scientific
Microscopes
Ultra-low focus control, angled viewing head, and interpupillary / dioptric adjustments make this microscope comfortable to use.
  • The compact frame allows easy arm placement to the controls
  • Plan objectives provide exceptionally high optical flatness and visual clarity
  • Choose from binocular or trinocular viewing heads, and Brightfield or Phase Contrast configurations
Optical system: VIOS infinity corrected.
Dioptric: −5 to +5
Ordering information: 5 year optical/mechanical; 1 year electrical warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty