Would you like to log in again?
VanGuard 1353SL Stereo Microscope, Vee Gee Scientific
Supplier: Vee Gee Scientific
Vanguard
1353SL
76545-994EA 587.38 USD InStock
76545-994
VanGuard 1353SL Stereo Microscope, Vee Gee Scientific
Microscopes
VanGuard 1353SL Stereo Microscope features precision optics to ensure the highest level of performance for inspection and dissection applications.
  • Upper and lower LED lighting provides bright and efficient illumination
  • Large stage area gives you room to work with bigger objects
  • LED illumination
Optical system: Fixed 160 mm
Dioptric: −5 to +5
Ordering information: 5 year optical/mechanical; 1 year electrical warranty.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty