Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Hydrometer Jars
Supplier: Heathrow Scientific
21050-600 21050-1000-L 21050-100 21050-1000 21050-500 21050-250
76546-094EA 26.74 USD InStock
76546-094 76546-092 76546-102 76546-100 76546-098 76546-096
Hydrometer Jars
Cylinders Hydrometer Cylinders
Hydrometer jars are made of borosilicate glass and feature a beaded rim with a circular base.
  • Available in a variety of capacities
All hydrometer jars are sized properly to eliminate surface tension effects that lead to inaccurate readings. Models 76546-096 have a graduation line at 1000 ml for precise filling and is designed for use as a sedimentation cylinder.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty