Would you like to log in again?
Vee Gee Volumetric Flasks with Snap Cap
Supplier: Heathrow Scientific
Vee Gee
2303A-500 2303A-100 2303A-10 2303A-50 2303A-250 2303A-25 2303A-1000 2303A-200
76546-118CS 4932.78 USD InStock
76546-118 76546-116 76546-114 76546-124 76546-112 76546-122 76546-110 76546-120
Vee Gee Volumetric Flasks with Snap Cap
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for snap-cap fit.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 68 °F (20 °C)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a snap cap
  • Wide range of sizes available (10 to 1000 ml)
Our volumetric flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions. Each flask is supplied with a snap cap which helps protect against breakage and spills.
Certifications: Meets ASTM E-288 Class A specifications and USP standards for glassware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty