Would you like to log in again?
Vee Gee Volumetric Flasks with Glass Stopper
Supplier: Heathrow Scientific
Vee Gee
2306A-250 2306A-50 2306A-5 2306A-200 2306A-10 2306A-2000 2306A-100 2306A-1000 2306A-25 202GK-004 2306A-500
490011-060CS 468.77 USD InStock
490011-060 76546-128 76546-138 76546-136 76546-146 76546-144 76546-132 76546-142 76546-130 76546-042 76546-140
Vee Gee Volumetric Flasks with Glass Stopper
Flasks Volumetric Flasks
These volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Wide range of sizes available (5 to 2000 ml)
Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions. Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
Certifications: Meets ASTM E-288 Class A specifications and USP standards for glassware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty