Would you like to log in again?
Vee Gee Volumetric Flasks with Glass Stoppers, Amber
Supplier: Heathrow Scientific
Vee Gee
2307A-25 2307A-200 2307A-50 2307A-1000 2307A-500 2307A-20 2307A-100 2307A-250 2307A-10
76546-150CS 3920.78 USD InStock
76546-150 76546-160 76546-148 76546-158 76546-156 76546-154 76546-164 76546-152 76546-162
Vee Gee Volumetric Flasks with Glass Stoppers, Amber
Flasks Volumetric Flasks
These volumetric flasks are constructed from low-actinic amber glass, providing protection for materials that are sensitive to light. 
  • Low-actinic amber glass for light sensitive materials
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Wide range of sizes available (10 to 1000 ml)
These flask feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers. Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
Certifications: Meets ASTM E-288 Class A specifications and USP standards for glassware.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty