Would you like to log in again?
Brix Hydrometers
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6601-10 6601-5 6601-4 6601-14 6601-13 6601-3 6601-15 6601-11 6601-1 6601-2
76546-282EA 31.1 USD InStock
76546-282 76546-270 76546-280 76546-278 76546-288 76546-276 76546-286 76546-274 76546-284 76546-272
Brix Hydrometers
Hydrometers
These brix scale hydrometers are calibrated to show the percentage of sucrose by weight at 20 °C. The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
  • Calibrated to show percent by weight
  • Wide variety of Brix ranges available
The data of the National Bureau of Standards are used.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty