Would you like to log in again?
Brix Hydrometers with Thermometer (°F)
Supplier: Heathrow Scientific
6601TS-10F 6601TS-2F 6601TS-1F 6601TS-13F 6601TS-3F
76546-306EA 53.09 USD InStock
76546-306 76546-314 76546-300 76546-310 76546-294
Brix Hydrometers with Thermometer (°F)
Hydrometers
Our brix scale hydrometers with ℉ thermometer are calibrated to show the percentage of sucrose by weight at 68 ℉.
  • Calibrated to show percent sucrose by weight
  • Wide variety of brix ranges available
  • Thermometer liquid fill is safety blue
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (68 ℉).
The thermometer scale is located in the body and contains safety blue liquid fill. The thermometers have a range of 30 to 150 ℉ with 2° subdivisions. A correction scale for conversion of reading to 68 ℉ is printed opposite the temperature scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty