Would you like to log in again?
Brix Hydrometers with Thermometer (°C)
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6601TS-1 6601TS-3 6601TS-15 6601TS-12 6601TS-14 6601TS-2 6601TS-10 6601TS-6 6601TS-4 6601TS-13 6601TS-8 6601TS-5
76546-308EA 53.09 USD InStock
76546-308 76546-292 76546-318 76546-290 76546-316 76546-304 76546-302 76546-312 76546-322 76546-320 76546-298 76546-296
Brix Hydrometers with Thermometer (°C)
Hydrometers
Our brix scale hydrometers with °C thermometer are calibrated to show the percentage of sucrose by weight at 20 °C.
  • Calibrated to show percent sucrose by weight
  • Wide variety of brix ranges available
  • Thermometers contain safety blue liquid fill
The Brix scale is based on the following: 1° Brix = 1% sucrose by weight at specified temperature (20 °C).
The thermometer scale is located in the body and features safety blue liquid fill. The thermometers have a range of 0 to 50 °C with 1° subdivisions. A correction scale for conversion of reading to 20 °C is printed opposite of the temperature scale.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty