Would you like to log in again?
Precision Short Form Specific Gravity Hydrometers
Supplier: Heathrow Scientific
6602-9S 6602-8S 6602-7S 6602-4S 6602-5S 6602-10S 6602-6S
76546-326EA 29.46 USD InStock
76546-326 76546-358 76546-346 76546-366 76546-354 76546-362 76546-350
Precision Short Form Specific Gravity Hydrometers
Hydrometers
These precision short form specific gravity hydrometers use less volume of the test liquid while still offering 0.001 resolution.
Each of these hydrometers has a small specific gravity range and has been calibrated for use at 60 °F.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty