Would you like to log in again?
Ultra Precision Specific Gravity Hydrometers
Supplier: Heathrow Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
6602-11 6602-15 6602-4 6602-6 6602-9 6602-7 6602-8 6602-5 6602-10 6602-2 6602-13 6602-12 6602-14 6602-3 6602-19
76546-328EA 30.96 USD InStock
76546-328 76546-338 76546-348 76546-336 76546-324 76546-356 76546-334 76546-344 76546-332 76546-364 76546-342 76546-352 76546-330 76546-340 76546-360
Ultra Precision Specific Gravity Hydrometers
Hydrometers
These ultra precision specific gravity hydrometers measure the ratio of the mass of a solid or liquid to the mass of an equal volume of distilled water.
These hydrometers feature smaller ranges but with higher resolution and have all been calibrated for use at 60 °F.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty